Anti-Melanoma Associated Antigen 100+ / 7 kDa Mouse Monoclonal Antibody [clone: NKI/beteb]

Lieferant: Abcam
ABCAAB34165-1EA 697 EUR
ABCAAB34165-1
Anti-Melanoma Associated Antigen 100+ / 7 kDa Mouse Monoclonal Antibody [clone: NKI/beteb]
Antikörper
Type: Primary
Antigen: Melanoma Associated Antigen 100+ / 7 kDa
Clonality: monoclonal
Clone: NKI/beteb
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR