Anti-NQO1 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB34173-100EA 719 EUR
ABCAAB34173-100
Anti-NQO1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: NQO1 (NAD(P)H dehydrogenase, quinone 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Guinea pig, Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR