Anti-ITGA1 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: TS2/7]
Lieferant: Abcam
ABCAAB34176-50EA 876 EUR
ABCAAB34176-50
Anti-ITGA1 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: TS2/7]
Antikörper
Type: Primary
Antigen: ITGA1 (integrin, alpha 1)
Clonality: monoclonal
Clone: TS2/7
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human