Anti-CD44v3 Mouse Monoclonal Antibody [clone: VFF-327]

Lieferant: Abcam
ABCAAB34229-50EA 926 EUR
ABCAAB34229-50
Anti-CD44v3 Mouse Monoclonal Antibody [clone: VFF-327]
Antikörper
Type: Primary
Antigen: CD44v3
Clonality: monoclonal
Clone: VFF-327
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR