Anti-Neurexin II alpha Rabbit Polyclonal Antibody
Lieferant: Abcam
ABCAAB34245-100EA 578 EUR
ABCAAB34245-100
Anti-Neurexin II alpha Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Neurexin II alpha
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat