Anti-MADCAM1 Mouse Monoclonal Antibody [clone: 314G8]

Lieferant: Abcam
ABCAAB34247-100EA 887 EUR
ABCAAB34247-100
Anti-MADCAM1 Mouse Monoclonal Antibody [clone: 314G8]
Antikörper
Type: Primary
Antigen: MADCAM1 (mucosal vascular addressin cell adhesion molecule 1)
Clonality: monoclonal
Clone: 314G8
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR