Anti-Fibrinogen Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB34269-125EA 713 EUR
ABCAAB34269-125
Anti-Fibrinogen Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Fibrinogen (fibrinogen)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR