Anti-IL4 Rat Monoclonal Antibody [clone: MP4-25D2]

Lieferant: Abcam
ABCAAB34277-100EA 672 EUR
ABCAAB34277-100
Anti-IL4 Rat Monoclonal Antibody [clone: MP4-25D2]
Antikörper
Type: Primary
Antigen: IL4 (interleukin 4)
Clonality: monoclonal
Clone: MP4-25D2
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR