Anti-CD7 Mouse Monoclonal Antibody (Biotin) [clone: LT7]

Lieferant: Abcam
ABCAAB34293-50EA 375 EUR
ABCAAB34293-50
Anti-CD7 Mouse Monoclonal Antibody (Biotin) [clone: LT7]
Antikörper
Type: Primary
Antigen: CD7 (CD7 molecule)
Clonality: monoclonal
Clone: LT7
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR