Anti-IL6ST Mouse Monoclonal Antibody [clone: B-R3]

Lieferant: Abcam
ABCAAB34315-100EA 890 EUR
ABCAAB34315-100
Anti-IL6ST Mouse Monoclonal Antibody [clone: B-R3]
Antikörper
Type: Primary
Antigen: IL6ST (interleukin 6 signal transducer (gp130, oncostatin M receptor))
Clonality: monoclonal
Clone: B-R3
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR