Anti-Choline Acetyltransferase Chicken Polyclonal Antibody
Lieferant: Abcam
ABCAAB34419-100EA 860 EUR
ABCAAB34419-100
Anti-Choline Acetyltransferase Chicken Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Choline Acetyltransferase
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human, Mouse, Rat