Anti-ITGA1 Mouse Monoclonal Antibody [clone: TS2/7]

Lieferant: Abcam
ABCAAB34445-100EA 682 EUR
ABCAAB34445-100
Anti-ITGA1 Mouse Monoclonal Antibody [clone: TS2/7]
Antikörper
Type: Primary
Antigen: ITGA1 (integrin, alpha 1)
Clonality: monoclonal
Clone: TS2/7
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR