Anti-AHSG Goat Polyclonal Antibody

Lieferant: Abcam
ABCAAB34505-100EA 666 EUR
ABCAAB34505-100
Anti-AHSG Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: AHSG (alpha-2-HS-glycoprotein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR