Anti-TOR1A Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB34540-100EA 713 EUR
ABCAAB34540-100
Anti-TOR1A Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: TOR1A (torsin family 1, member A (torsin A))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR