Anti-Hepatitis B Virus Surface Antigen (Ad/Ay) Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: Abcam
ABCAAB34553-500EA 666 EUR
ABCAAB34553-500
Anti-Hepatitis B Virus Surface Antigen (Ad/Ay) Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
Type: Primary
Antigen: Hepatitis B Virus Surface Antigen (Ad/Ay)
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR