Anti-HSV1 + HSV2 Goat Polyclonal Antibody

Lieferant: Abcam
ABCAAB34568-500EA 666 EUR
ABCAAB34568-500
Anti-HSV1 + HSV2 Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: HSV1 + HSV2
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR