Anti-GST Mouse Monoclonal Antibody [clone: 3D4]

Lieferant: Abcam
ABCAAB34589-100EA 882 EUR
ABCAAB34589-100
Anti-GST Mouse Monoclonal Antibody [clone: 3D4]
Antikörper
Type: Primary
Antigen: GST
Clonality: monoclonal
Clone: 3D4
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR