Anti-Lefty Mouse Monoclonal Antibody [clone: 7C5G1H6H10]

Lieferant: Abcam
ABCAAB34593-100EA 703 EUR
ABCAAB34593-100
Anti-Lefty Mouse Monoclonal Antibody [clone: 7C5G1H6H10]
Antikörper
Type: Primary
Antigen: Lefty
Clonality: monoclonal
Clone: 7C5G1H6H10
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR