Anti-KLH Mouse Monoclonal Antibody [clone: 12B4.G3.A8]

Lieferant: Abcam
ABCAAB34607-100EA 697 EUR
ABCAAB34607-100
Anti-KLH Mouse Monoclonal Antibody [clone: 12B4.G3.A8]
Antikörper
Type: Primary
Antigen: KLH (Keyhole Limpet Hemocyanin)
Clonality: monoclonal
Clone: 12B4.G3.A8
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR