Anti-ERBB3 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB34641-250EA 672 EUR
ABCAAB34641-250
Anti-ERBB3 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ERBB3 (v-erb-b2 avian erythroblastic leukemia viral oncogene homolog 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR