Anti-MIF Chicken Polyclonal Antibody

Lieferant: Abcam
ABCAAB34644-250EA 672 EUR
ABCAAB34644-250
Anti-MIF Chicken Polyclonal Antibody
Antikörper
Type: Primary
Antigen: MIF (macrophage migration inhibitory factor (glycosylation-inhibiting factor))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR