Anti-SUMO3 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB34661-500EA 703 EUR
ABCAAB34661-500
Anti-SUMO3 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SUMO3 (small ubiquitin-like modifier 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR