Anti-IL1RL2 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB34665-200EA 666 EUR
ABCAAB34665-200
Anti-IL1RL2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: IL1RL2 (interleukin 1 receptor-like 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR