Anti-CAMK2B Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB34703-100EA 713 EUR
ABCAAB34703-100
Anti-CAMK2B Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CAMK2B (calcium/calmodulin-dependent protein kinase II beta)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR