Anti-E1 Ubiquitin Activating Enzyme Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB34711-500EA 688 EUR
ABCAAB34711-500
Anti-E1 Ubiquitin Activating Enzyme Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: E1 Ubiquitin Activating Enzyme
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR