Anti-R Phycoerythrin Goat Polyclonal Antibody (TRITC (Rhodamine))

Lieferant: Abcam
ABCAAB34717-100EA 666 EUR
ABCAAB34717-100
Anti-R Phycoerythrin Goat Polyclonal Antibody (TRITC (Rhodamine))
Antikörper
Type: Primary
Antigen: R Phycoerythrin
Clonality: polyclonal
Clone:
Conjugation: TRITC (Rhodamine)
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR