Anti-KDM5C Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB34718-200EA 794 EUR
ABCAAB34718-200
Anti-KDM5C Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: KDM5C (lysine (K)-specific demethylase 5C)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR