Anti-TNF Rat Monoclonal Antibody [clone: MP6-XT22]

Lieferant: Abcam
ABCAAB34719-100EA 666 EUR
ABCAAB34719-100
Anti-TNF Rat Monoclonal Antibody [clone: MP6-XT22]
Antikörper
Type: Primary
Antigen: TNF (tumor necrosis factor)
Clonality: monoclonal
Clone: MP6-XT22
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR