Anti-R Phycoerythrin Goat Polyclonal Antibody

Lieferant: Abcam
ABCAAB34721-100EA 666 EUR
ABCAAB34721-100
Anti-R Phycoerythrin Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: R Phycoerythrin
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR