Anti-VDAC1 Rabbit Polyclonal Antibody
Lieferant: Abcam
ABCAAB34726-100EA 906 EUR
ABCAAB34726-100
Anti-VDAC1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: VDAC1 (voltage-dependent anion channel 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat