Anti-GST Goat Polyclonal Antibody (AP (Alkaline Phosphatase))

Lieferant: Abcam
ABCAAB34728-500EA 690 EUR
ABCAAB34728-500
Anti-GST Goat Polyclonal Antibody (AP (Alkaline Phosphatase))
Antikörper
Type: Primary
Antigen: GST
Clonality: polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR