Anti-R Phycoerythrin Goat Polyclonal Antibody (Biotin)

Lieferant: Abcam
ABCAAB34730-250EA 666 EUR
ABCAAB34730-250
Anti-R Phycoerythrin Goat Polyclonal Antibody (Biotin)
Antikörper
Type: Primary
Antigen: R Phycoerythrin
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR