Anti-HP Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB34732-250EA 690 EUR
ABCAAB34732-250
Anti-HP Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: HP (haptoglobin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR