Anti-GST Goat Polyclonal Antibody (Texas Red®)

Lieferant: Abcam
ABCAAB34733-500EA 666 EUR
ABCAAB34733-500
Anti-GST Goat Polyclonal Antibody (Texas Red®)
Antikörper
Type: Primary
Antigen: GST
Clonality: polyclonal
Clone:
Conjugation: Texas Red®
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR