Anti-Rubella Virus capsid Mouse Monoclonal Antibody

Lieferant: Abcam
ABCAAB34749-100EA 688 EUR
ABCAAB34749-100
Anti-Rubella Virus capsid Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: Rubella Virus capsid
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR