Anti-Parainfluenza Virus type 1 nucleocapsid Mouse Monoclonal Antibody

Lieferant: Abcam
ABCAAB34752-100EA 649 EUR
ABCAAB34752-100
Anti-Parainfluenza Virus type 1 nucleocapsid Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: Parainfluenza Virus type 1 nucleocapsid
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR