Anti-Hantavirus nucleocapsid protein Mouse monoclonal antibody

Lieferant: Abcam
ABCAAB34759-1EA 590 EUR
ABCAAB34759-1
Anti-Hantavirus nucleocapsid protein Mouse monoclonal antibody
Type: Primary
Antigen: Hantavirus nucleocapsid protein
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR