Anti-NOXO1 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB34761-100EA 690 EUR
ABCAAB34761-100
Anti-NOXO1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: NOXO1 (NADPH oxidase organizer 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR