Anti-Hantavirus glycoprotein G2 Mouse Monoclonal Antibody
Lieferant: Abcam
ABCAAB34763-100EA 882 EUR
ABCAAB34763-100
Anti-Hantavirus glycoprotein G2 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: Hantavirus glycoprotein G2
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen