Anti-TRIM27 Rabbit Polyclonal Antibody (Biotin)

Lieferant: Abcam
ABCAAB34771-100EA 719 EUR
ABCAAB34771-100
Anti-TRIM27 Rabbit Polyclonal Antibody (Biotin)
Antikörper
Type: Primary
Antigen: TRIM27 (tripartite motif containing 27)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR