Anti-Myc Tag Rabbit Polyclonal Antibody (Biotin)

Lieferant: Abcam
ABCAAB34773-100EA 697 EUR
ABCAAB34773-100
Anti-Myc Tag Rabbit Polyclonal Antibody (Biotin)
Antikörper
Type: Primary
Antigen: Myc Tag (Myc tag)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR