Anti-Sts1 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB34781-100EA 666 EUR
ABCAAB34781-100
Anti-Sts1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Sts1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR