Anti-NPFF1 Receptor Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB34785-100EA 666 EUR
ABCAAB34785-100
Anti-NPFF1 Receptor Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: NPFF1 Receptor
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR