Anti-Glutamate Dehydrogenase Rabbit polyclonal antibody unconjugated

Lieferant: Abcam
ABCAAB34786-500EA 713 EUR
ABCAAB34786-500
Anti-Glutamate Dehydrogenase Rabbit polyclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: Glutamate Dehydrogenase
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR