Anti-NPFF2 Receptor Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB34787-100EA 634 EUR
ABCAAB34787-100
Anti-NPFF2 Receptor Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: NPFF2 Receptor
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR