Anti-KIDINS220 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB34792-100EA 697 EUR
ABCAAB34792-100
Anti-KIDINS220 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: KIDINS220 (kinase D-interacting substrate, 220kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR