Anti-Avidin Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB34824-500EA 2800 EUR
ABCAAB34824-500
Anti-Avidin Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Avidin
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Chicken
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR