Anti-GPT Sheep Polyclonal Antibody

Lieferant: Abcam
ABCAAB34832-500EA 666 EUR
ABCAAB34832-500
Anti-GPT Sheep Polyclonal Antibody
Antikörper
Type: Primary
Antigen: GPT (glutamic-pyruvate transaminase (alanine aminotransferase))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Pig
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR