Anti-Red Blood Cells Rabbit Polyclonal Antibody
Lieferant: Abcam
ABCAAB34858-500EA 882 EUR
ABCAAB34858-500
Anti-Red Blood Cells Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Red Blood Cells
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human