Anti-RNF7 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB34873-100EA 734 EUR
ABCAAB34873-100
Anti-RNF7 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: RNF7 (ring finger protein 7)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR